VWR® −40 °C Ultra-Low Temperature Freezers
Supplier: VWR International
76307-984 76307-990 VWR60040FA
76307-984EA 21323.17 CAD
76307-984 76307-990 77661-482
VWR® −40 °C Ultra-Low Temperature Freezers
Freezers Ultra Low Temperature Freezers
Dependable –40 °C storage, powered by green, hydrocarbon refrigerants delivering operational savings and sustainability.
  • Low energy
  • Low peak variation
  • Fast door opening recovery
  • Long warm up time
  • Three sizes
VWR -40 °C ULT freezers are built for seamless operation with user-friendly features. With low noise operation, flexible rack configurations and faster re-entry times, these -40 °C ULT freezers are designed to enhance the overall user experience and meet the needs of busy laboratories.
Certifications:
cULus, SNAP, CE marked
Ordering information: Options and accessories are sold separately; contact your VWR representative for more information.
Caution:
Must be removed from pallet upon delivery. Fits through standard 34.5" doorways.