Cage-Klenz® 220 Acid-Based Detergents
Supplier: Steris
Cage-Klenz®
1K22D3 1K2205 1K2202
77531-228EA 15996.54 CAD
77531-228 14004-182 14004-184
Cage-Klenz® 220 Acid-Based Detergents
Cleaning Agents Detergents and Surface Cleansers
Cage-Klenz® 220 Detergent is a liquid phosphate-free, hydroxyacetic acid-based detergent.
  • Efficacy - Effectively cleans hard water and urine scale, and organic soils in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Cage-Klenz® 220 Detergent is specially formulated for the safe and effective removal of hard water scale, urine scale and organic residues found in research animal caging.