Cage-Klenz® 200 Acid-Based Detergents
Supplier: Steris
1K2002 1K2005 1K20D9
CA14004-124EA 2595.43 CAD
CA14004-124 CA14223-964 77531-222
Cage-Klenz® 200 Acid-Based Detergents
Cleaning Agents Detergents and Surface Cleansers
Cage-Klenz® 200 Detergent is a phosphoric acid-based detergent for use in high pressure spray animal cage washing equipment.
  • Efficacy - Effectively cleans hard water and urine scale, and metal oxides in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Cage-Klenz® 200 Detergent is specially formulated to remove challenging hard water and urine scales found in research animal caging.