Cage-Klenz® 100 Alkaline Detergents
Supplier: Steris
1K1002 1K1005 1K10D9
CA14004-188EA 2287.54 CAD
CA14004-188 CA21899-716 77531-218
Cage-Klenz® 100 Alkaline Detergents
Cleaning Agents Detergents and Surface Cleansers
Cage-Klenz® 100 Detergent is a liquid, phosphate-free detergent for use in high pressure spray animal cage washing equipment. It is formulated with corrosion inhibitors for enhanced materials compatibility.
  • Efficacy - Effectively cleans organic cage residues in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Cage-Klenz® 100 Detergent is specially formulated to meet the unique cleaning and materials compatibility requirements found in research animal caging.