Overpack and Salvage Drums, UN and DOT, Eagle Mfg
Supplier: Eagle Manufacturing
1665 1695 1602 1654
56617-337EA 1403.07 CAD
56617-337 56617-339 56618-528 56618-550
Overpack and Salvage Drums, UN and DOT, Eagle Mfg
Barrels, Kegs, Drums
Designed for use with hazardous materials, Packing Groups I, II, and III.
  • Constructed of blow-molded high-density polyethylene with UV inhibitors for superior strength
  • Chemically-resistant to most materials
  • Safety yellow with removable head
  • Forkliftable
  • Nestable for easy storage
  • Suitable for secondary containment storage, transportation, and cleanup
Salvage drums have a metal bolt ring locking band and meet 3psi pressure test for Salvage Drum Compliance. Overpack drums have a metal locking band. All drums are UN certified to HM-181. Salvage drums meetMore Product Information