Cage-Klenz® 250 Acid-Based Detergents
Supplier: Steris
Cage-Klenz®
1K2505 1K2505 1K25D9 1K25-02 1K2502
CA14004-180EA 2330.46 CAD
14004-180 CA14004-180 CA14004-178 77531-214 14004-178
Cage-Klenz® 250 Acid-Based Detergents
Cleaning Agents Detergents and Surface Cleansers
Cage-Klenz® 250 Detergent is a liquid phosphate-free, citric acid-based detergent.
  • Efficacy - Effectively cleans hard water and urine scale in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Cage-Klenz® 250 Detergent is specially formulated for the safe and effective removal of hard water scale, urine scale and organic residues found in research animal caging.