AlphaTec® 53-003 Broad Chemical Protection Gloves with Nylon Liner
Supplier: Ansell Healthcare
53003070 53003080 53003090 53003110
77528-923CS 1597.02 CAD
77528-923 77528-929 77528-915 77528-907
AlphaTec® 53-003 Broad Chemical Protection Gloves with Nylon Liner
Gloves
Revolutionary Hybrid Technology delivering a breakthrough in chemical performance.
  • MICROCHEM™ multi-layer barrier features unique rubber polymer and plastic laminate technology
  • Designed to provide extreme protection against a wide range of chemical types
  • Contact heat approved up to 100 °C/212 °F
  • Anti-static according to EN 1149-3 standards
  • Made without natural rubber or silicone, ensuring protection against mechanical risks and allergic reactions
  • Unique manufacturing process and nylon liner to enhance comfort during use
The AlphaTec® 53-003 series are revolutionary industrial gloves that redefine chemical protection. The innovative design combines an advanced rubber polymer coating and plastic laminate technology improving chemical permeation while delivering comfort and tactility. These safety gloves meet the stringent requirements of EN ISO 374:2016, with 17 out of 18 candidate chemicals achieving a minimum Class 2 classification, and 16 out of 17 achieving class 6>480 minutes. In addition to chemical protection, they satisfy EN 407 contact heat level 1, preventing workplace injuries. They also possess EN-compliant abrasion resistance.
The unique manufacturing process allows for the glove to be form fitting, offering elevated comfort and dexterity.
Certifications: CE 0493; EN 421:2010; EN ISO 374-5:2016; EN 388:2016+A1:2018; EN407:2020.
Packaging:
1 pair per bag, 36 pairs per carton.