Vee Gee Separatory Funnels
Supplier: Heathrow Scientific
1426T-1000 1426T-2000 1426T-300 1426T-500
76546-006EA 285.06 CAD
76546-006 76546-008 76546-010 76546-012
Vee Gee Separatory Funnels
Funnels Separating Funnels
Our separatory funnels are an excellent tool for liquid-liquid extractions, phase separations, and chemical processes in laboratories.
  • Supplied with PTFE stopcocks to eliminate freezing and lubricant contamination
  • Made from high-quality borosilicate tubing with a low coefficient of expansion
  • Features smooth tooled tips, white block letters, and large marking squares
  • Squibb style
Whether you're in a research laboratory, educational institution, or industrial setting, our separatory funnels empower you to achieve the highest standards of accuracy in your separation procedures.
These separatory funnels feature smooth tooled tips, large white block letters, and large marking squares. Each funnel is made from borosilicate tubing with a low coefficient of expansion, and are supplied with PTFE stopcocks, which eliminates freezing and lubricant contamination.