Vee Gee Glass Stoppers
Supplier: Heathrow Scientific
03081 03082 03083 03084 03085 03087
76545-840PK 36.82 CAD
76545-840 76545-842 76545-844 76545-846 76545-848 76545-850
Vee Gee Glass Stoppers
Stoppers Glass Stoppers
These stoppers are lightweight yet very strong to reduce chipping and breakage, thus minimizing replacement costs.
  • Meet ASTM E-675 specifications
  • Flat top design allows stopper to stand on its head
  • Fully interchangeable with other common stoppers
  • Clear color
  • Comes in packs of 6
The flat top design allows the stopper to stand on its head, which frees the user’s hand and minimizes potential contamination. They are fully interchangeable with other common flask stoppers and fit all standard labware of comparable size.