LabKlenz® Glassware Washing Detergents, STERIS®
Supplier: Steris
1L2502 1L2508
CA89089-214EA 2010.39 CAD
CA89089-214 CA89089-218
LabKlenz® Glassware Washing Detergents, STERIS®
Cleaning Agents Detergents and Surface Cleansers
Ideal for removing residues from laboratory glassware in high-energy automated washing applications, manual cleaning, ultrasonic cleaning, and component parts cleaning
  • Free-rinsing and low-foaming across a wide temperature range
  • Easily removed from surfaces to prevent residues
  • Contain biodegradable surfactants. Environmentally friendly.
LabKlenz® 100 detergent is a phosphate-free, liquid alkaline detergent with corrosion inhibitors for enhanced substrate compatibility. Extremely free-rinsing to meet critical laboratory requirements. pH (1%): 11.5 (typical).
LabKlenz® 110 detergent is a phosphate-free, liquid alkaline detergent that is extremely free-rinsing to meet critical laboratory requirements. pH (1%): 12.4 (typical).
LabKlenz® 120 detergent is a mild alkaline liquid detergent that is compatible with a variety of substrates including stainless steel, mild steel, copper, and aluminum. Extremely free-rinsing to meet critical laboratory requirements. pH (concentrate): 9.4 (typical).
LabKlenz® 200 detergent is phosphoric acid-based and formulated for the removal of scale, particulate, and inorganic soils. pH (1%): 2.0 (typical).
LabKlenz® 250 detergent is citric acid-based and formulated for use as a rinse aid, as well as for the removal of particulate, scale, and inorganic soils. pH (1%): 2.6 (typical).
Certifications: Manufactured in an FDA-registered and ISO 9001/13485-certified facility.