Tergajet® Low-Foaming Powdered Detergents
Supplier: Alconox
2204 2201
CA80077-368CS 732 CAD
CA80077-368 CA80077-374
Tergajet® Low-Foaming Powdered Detergents
Cleaning Agents Detergents and Surface Cleansers
Tergajet® is a nonionic, low foaming phosphate free powdered cleaner for use in labware washers, manual and ultrasonic cleaning.
  • Passes inhibitory residue test Dilute 1:200
  • Concentrated
  • Phosphate-free, biodegradable and readily disposable
  • Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
  • Low foaming heavy duty detergent for high performance results
  • Free rinsing to give you reliable results and no interfering residues
Ideal for under-counter and floor standing lab washing machines. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass. Free rinsing for reliable results and no interfering residues.
Used to remove soil, radioisotopes, radioactivity, organics, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, particulates, inorganic residues, chemical and solvents. Recommended for glass, stainless steel, PTFE, ceramic, cement and alkali resistant plastics and metals. Tergajet is inhibited for aluminum.
Used to clean water testing labware cleaning, phosphate sensitive lab equipment cleaning, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors USDA food contact surface cleaning compliant.
Cleaning method: Laboratory warewashers, dishwashers, washer-sterilizers, washersanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners, clean-inplace and manual cleaning if proper skin and eye protection is used.
Note: Corrosion testing is advisable.
Certifications: FDA certified.