Complete II One-Step Organic Acid Cleaner/Descaler, CANI
Supplier: Cani
CAN-COM2-15 CAN-COM2-275 CAN-COM2-30 CAN-COM2-55 CAN-COM2-5 CAN-COM2-2.5
CA93001-206EA 4189.23 CAD
CA93001-206 CA93001-212 CA93001-214 CA93001-216 CA93001-218 CA93001-210
Complete II One-Step Organic Acid Cleaner/Descaler, CANI
Cleaning Agents Detergents and Surface Cleansers
Glycolic/citrus acid cleaner/descaler formulated for the research, pharmaceutical, and biotech industries
Cleaner is ideal for animal cage wash applications and will clean and descale in just one wash cycle, eliminating the use of an alkaline detergent and a full wash cycle, improving throughput, and reducing water consumption. Effective in pharmaceutical/biotech process cleaning and derouging. Features a low-foaming formulation that will not cavitate recirculation pumps. Cleaner’s free-rinsing design provides a clean, residue-free surface, and the excellent hard water toleranceMore Product Information