Waste Gas Control Supplies, VetEquip®
Supplier: Vetequip
931435 931436 931440 931401 931443
89012-610EA 8.12 CAD
89012-610 89012-612 89012-614 89012-608 89012-616
Waste Gas Control Supplies, VetEquip®
Animal Supplies Animal Anesthesia Systems
The scavenging interface kit features an interface valve that is installed between an anesthesia machine and a vacuum evacuation system. The kit helps control and monitor exposure to anesthetic waste gases.
  • Valve equalizes both positive and negative pressure so that no metabolic gases are pulled past the patient
  • Monitoring badge adsorbs halogenated hydrocarbons, including isoflurane and halothene
  • After <=8 hours of exposure, the badge should be sent to a chemical laboratory for analysis of exposure levels
The activated charcoal adsorption filter adsorbs passively and has been proven effective in eliminating waste anesthetic gas pollution.
Ordering information: Kit includes a manufacturer’s one-year money-back guarantee and seven-year replacement warranty, one interface valve, one 3 l (0.8 gal.) reservoir bag, 1.8 m (6') of 6 mm (¼") I.D. suction hose, and 1.8 m (6') of 19 mm (¾") evacuation tubing.