CP-712 Acid Neutralizer, CANI
Supplier: Cani
CAN-712-15 CAN-712-275 CAN-712-30 CAN-712-55 CAN-712-5 CAN-712-2.5
CA93001-168EA 4502.51 CAD
CA93001-168 CA93001-172 CA93001-174 CA93001-176 CA93001-178 CA93001-170
CP-712 Acid Neutralizer, CANI
Cleaning Agents Detergents and Surface Cleansers
Phosphate-free, concentrated liquid alkaline cleaning agent formulated to be used after the application of acid cleaners for the pharmaceutical, biotechnology, cosmetic, and medical device industries
Effectively removes a wide range of process and research residues, including serum, proteins, organics, and oils. Cleaning agent works as an excellent laboratory animal cage wash product at 0.2–1% use dilution. Designed to be low foaming at all temperatures for easy rinsing and will not cavitate recirculation pumps. Free-rinsing formulation provides clean, residue free surfaces. ChelantsMore Product Information