VWR® Support Jacks, Stainless Steel
Supplier: VWR
VWR®
960022 960027 960025 960021 960026 960023 960024
14233-360EA 377.93 CAD
14233-360 14233-370 14233-362 14233-372 14233-364 14233-366 14233-368
VWR® Support Jacks, Stainless Steel
Laboratory Jacks
Stainless steel lab jacks are ideal for use in fume hoods or benchtops.
  • Stainless steel construction
  • Autoclavable and chemical-resistant
  • Seven convenient sizes
These ultra-stable lifting platforms hold a variety of items such as glassware, hot plates, baths, and magnetic stirrers
Ratchet tool is designed to add extra leverage to the 30.5x30.5 cm (12x12") and 40.6x40.6 cm (16x16") support jacks. The ratcheting handle easily attaches to the actuating rod to allow easy, accurate adjustments.
Support rod kit includes a 43.2 cm (17") vertical support rod for mounting to the upper deck of a 15.2x15.2 cm (6x6"), 20.3x20.3 cm (8x8"), 25.4x25.4 cm (10x10"), 30.5x30.5 cm (12x12"), or 40.6x40.6 cm (16x16") support jack. Ideal for creating an adjustable support stand for mounting various items such as thermometer clamps, temperature probes, or flask and column clamps. Kit also includes a jam nut and a flat washer.
Constructed of 300-series stainless steel for exceptional strength and durability, support jacks are designed for use in extreme environments and high load applications. Durable construction allows jacks to be autoclaved or chemically cleaned. Support jacks are equipped with oversized, sure-grip adjustment knobs that provide extra leverage for easy height adjustments. Lab jacks are designed to hold the specified weight after it is lifted to the preferred height.