CP-700 Alkaline Detergent, CANI
Supplier: Cani
CAN-700-15 CAN-700-275 CAN-700-30 CAN-700-55 CAN-700-5 CAN-700-1 CAN-700-2.5
CA93001-154EA 4263.35 CAD
CA93001-154 CA93001-158 CA93001-160 CA93001-162 CA93001-164 CA93001-152 CA93001-156
CP-700 Alkaline Detergent, CANI
Cleaning Agents Detergents and Surface Cleansers
Low-foaming, concentrated liquid alkaline detergent used to clean process and research equipment, and laboratory glassware and plastics
Ideal for pharmaceutical, biotechnology, cosmetic, medical device, and food and beverage facilities. Detergent removes a wide range of process and product residues, including proteins, organics, many excipients, and fine chemicals. Detergent is free-rinsing and phosphate-free.
Order Now
SPECIFICATIONS
Environmentally Preferable