Serum Bottles, Borosilicate Glass, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
223738 223739 223742 223743 223745 223747 223746 223748 223760 223761 223762 223763 223764 223766 W223712
16171-308CS 456.6 CAD
16171-308 16171-319 16171-320 16171-330 16171-341 16171-352 16171-374 16171-385 16171-401 16171-433 16171-466 16171-499 16171-524 16171-557 89422-806
Serum Bottles, Borosilicate Glass, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Serum bottles are ideal for long and short term sample storage, lyophilization, and as vaccine/injectable drug containers.
  • Bottles fit most lyophilization applications
Clear bottles are manufactured from Wheaton 400 borosilicate molded glass, and conform to USP Type I requirements. Amber bottles are manufactured from Wheaton 500 borosilicate glass, and conform to USP Type I requirements. All bottles feature a 13Int.Ø×20Ext.Ø mm mouth opening, with the exception of 89422-806 which is 7Int.Ø×13Ext.Ø mm.
Ordering information: Bottles are packaged inMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable