Induction Chambers, VetEquip®
Supplier: VetEquip
941448 941443 941444 941454
89012-688EA 952.06 CAD
89012-688 89012-694 89012-686 89012-696
Induction Chambers, VetEquip®
Animal Supplies Animal Anesthesia Systems
Durable, clear acrylic VetEquip® chambers confine animals while allowing anesthetic gas to be circulated throughout the chamber.
  • Positive sealing gasket to help prevent gas leakage
  • Large easy-to-use latches
  • Standard anesthesia size inlet and outlet ports
  • Non-skid feet
  • 2 L, 7 L, and 15 L chambers can also be used for multiple animals.
1 L (0.3 gal.) chamber can hold a single mouse or small rat
2 L (0.5 gal.) chamber can hold several mice or a rat <=650 g (22.9 oz.)
7 L (1.8 gal.) chamber can hold extra-large rats, ferrets, and small to medium rabbits
15 L (4 gal.) chamber can hold large rabbits, cats, and very small non-human primates
Ordering information: Induction chambers include a manufacturer’s one-year money-back guarantee and seven-year replacement warranty.