Complete IV One-Step Phosphoric/Citric Acid Cleaner/Descaler, CANI
Supplier: Cani
CAN-COM4-15 CAN-COM4-275 CAN-COM4-30 CAN-COM4-55 CAN-COM4-5 CAN-COM4-2.5
CA93001-138EA 5172.81 CAD
CA93001-138 CA93001-144 CA93001-146 CA93001-148 CA93001-150 CA93001-140 CA93001-142
Complete IV One-Step Phosphoric/Citric Acid Cleaner/Descaler, CANI
Cleaning Agents Detergents and Surface Cleansers
Formulated for the research, pharmaceutical, and biotech industries
Cleaner is ideal for animal cage wash applications and will clean and descale in just one wash cycle, eliminating the use of an alkaline detergent and a full wash cycle, improving throughput, and reducing water consumption. Cleaner is also effective in pharmaceutical/biotech process cleaning and derouging. Features a low-foam formulation that will not cavitate recirculation pumps. Cleaner’s free-rinsing design provides a clean, residue-free surface, and the excellent hard water tolerance eliminates mineral deposits. TheMore Product Information