KIMAX® Viscometer Tube, Cannon-Fenske, Serialized, Uncalibrated, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
46460-100 46460-150 46460-350 46460-300 46460-50 46460-400 46460-200
52946-640EA 709.54 CAD
52946-640 52946-683 52946-661 52946-708 52946-720 52946-628 52946-606
KIMAX® Viscometer Tube, Cannon-Fenske, Serialized, Uncalibrated, Kimble Chase, DWK Life Sciences
Viscometers Capillary Viscometers
For obtaining kinematic viscosities of transparent liquids (ASTM Method of Test D445)
Designed from ASTM Specification D446. Permanently marked with an individual serial number. Lines and legend are printed black. Viscosity ranges shown below are for an efflux time greater than 200 seconds. Require calibration against a standard liquid of known viscosity or against a second viscometer with known constant.