Gas Supply Quick Disconnects, VetEquip®
Supplier: Vetequip
931506 931507 931509 931510 931511 931512 931513 931514 931515 931516 931517
89012-624EA 321.06 CAD
89012-624 89012-626 89012-630 89012-632 89012-634 89012-636 89012-638 89012-640 89012-642 89012-644 89012-646
Gas Supply Quick Disconnects, VetEquip®
Animal Supplies Animal Anesthesia Systems
The female end has a check valve and is usually installed on the hose connected to the gas supply, the male end is then installed on the hose connected to the machine.
  • Quick disconnects provide a fast and easy method for moving a machine without turning the gas supply off
  • Can be installed anywhere in the hose between the machine and the gas supply
Includes a manufacturer’s one-year money-back guarantee and seven-year replacement warranty.
Order Now
SPECIFICATIONS
Environmentally Preferable