CP-450 Concentrated Phosphoric Acid/Citric Cleaner and Descaler, CANI
Supplier: Cani
CAN-450-15 CAN-450-275 CAN-450-30 CAN-450-5 CAN-450-55 CAN-450-2.5
CA93001-264EA 4738.09 CAD
CA93001-264 CA93001-268 CA93001-270 CA93001-272 CA93001-274 CA93001-262 CA93001-266
CP-450 Concentrated Phosphoric Acid/Citric Cleaner and Descaler, CANI
Cleaning Agents Detergents and Surface Cleansers
Concentrated liquid formulated for the pharmaceutical, cosmetic, medical device, and food and beverage industries
Effectively removes a wide range of process and research residues, from inorganic salts and scales to particulate carbon and urine scales. Ideal for antacid equipment, animal cage wash applications, laboratory glassware washing, drug/biotech process cleaning, and as a final wash agent prior to pure water rinse. Routine use minimizes rust and rouge. Safe for use on both glassware and stainless steel substrates and a wide variety of plastics. Cleaner is designed to be low-foaming at all temperatures for easy rinsing and will not cavitate recirculation pumps. Free-rinsing formulation promotes a clean, residue-free surface. The inherent chelation of citric acid gives excellent hard water tolerance to eliminate mineral deposits.