Synthware Vacuum Adapters with Glass Stopcock, Kemtech America
Supplier: Kemtech America
A611420 A611922 A612440
60000-252EA 106.13 CAD
60000-252 60000-254 60000-256
Synthware Vacuum Adapters with Glass Stopcock, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Use this vacuum adapter with glass stopcock for regulating the flow of gases or liquids into reaction systems.
  • [ST] joint
  • Glass stopcock
  • With hose connection
Featuring a stopcock bore of 2 mm. Hose connection size of 8 mm O.D. at the middle serration.