Synthware Outer Joint Adapters with PTFE Stopcock, Kemtech America
Supplier: Kemtech America
A711420 A711922 A712440
60000-236EA 133.28 CAD
60000-236 60000-238 60000-240
Synthware Outer Joint Adapters with PTFE Stopcock, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Use this outer joint adapter with stopcock for regulating the flow of gases or liquids into reaction systems.
  • Outer [ST] joint
  • PTFE stopcock
  • With hose connection
Featuring a stopcock bore of 2 mm. Hose connection size is 8 mm ext. Ø. at the middle serration. With outer [ST] joint.