Synthware Vacuum/Argon/Nitrogen Adapters, Kemtech America
Supplier: Kemtech America
A301420 A301922 A302440 A302942
60000-172EA 40.88 CAD
60000-172 60000-174 60000-176 60000-178
Synthware Vacuum/Argon/Nitrogen Adapters, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Use this adapter for vacuum/Argon/Nitrogen applications.
  • Top hose connection
  • Bottom [ST] inner joint
Featuring an 8 mm hose connection on the top and male joint on the bottom.