Wall-Mounted Anesthesia Machines, VetEquip®
Supplier: Vetequip
901802 901803
89012-484EA 4593.15 CAD
89012-484 89012-486
Wall-Mounted Anesthesia Machines, VetEquip®
Animal Supplies Animal Anesthesia Systems
These wall-mounted machines are ideal for areas with limited floor and counter space.
  • Machines can be connected to an E-cylinder, H-tank, or in-house gas supply
  • The aluminum frame has a baked enamel finish that resists scratching and staining
The CO2 models have a CO2 absorber, which allows gas from the machine to circulate through absorbent material to remove CO2, then returns the anesthetic gas back to the patient.
Mounting bracket width: 406 mm (16")
Ordering information: The anesthesia machines are supplied with a manufacturer’s one-More Product Information