DECON-CLEAN®, Veltek Associates
Supplier: Veltek
DECON-Clean®
DC-04-1Z DC-01 DC-03-4Z DC-06-16Z-01 DC-05-1Z DC-02 DCWFI-SP-11Z
14003-290CS 946.17 CAD
14003-290 14003-292 14003-698 75795-366 14003-696 14003-694 14003-294
DECON-CLEAN®, Veltek Associates
Cleaning Agents Detergents and Surface Cleansers
Ideal for washable, nonporous surfaces such as walls, ceilings, floors, and stainless steel items, this cleaning agent is designed to remove residues left behind by disinfecting agents
DECON-Clean® is filtered at 0.2µm, gamma irradiated, and double-bagged. SimpleMix® one-step residue remover dilutes to 1:128 in Water for Injection. Sterile options are USP tested and delivered with a lot-specific certificate of irradiation and certificate of analysis. Traceable sterile cleaning agent comes with an LAL testing analysis and sterility report.