DECON-CYCLE II, Low pH Penolic, Veltek Associates
Supplier: Veltek Associates
DCY2-03-2Z DCY2-04-1/2Z DCY2-04-1Z DCY2-05-1/2Z DCY2-03-1Z
14004-470CS 547.85 CAD
14004-470 14222-696 75795-384 75795-386 76326-566
DECON-CYCLE II, Low pH Penolic, Veltek Associates
Cleaning Agents Detergents and Surface Cleansers
Phosphate-free detergent with multi-phenolic formula designed to clean, disinfect, and deodorize washable surfaces and kill a broad spectrum of pathogenic bacteria.
  • Filtered at 0.2 Microns
  • Double-bagged packaged
  • Completely traceable
Detergent features the SimpleMIX® system that ensures exact formulation of 1:256 in WFI water. It is validated for sterility and shelf life. Lot-specific certificates of analysis and sterility are also provided.
Recommended for use in pharmaceutical facilities, biotechnology facilities, medical device manufacturing, hospitals, and other healthcare institutions.