CIP 100® Alkaline Process and Research Cleaners
Supplier: Steris
1D1008 1D1001 1D1005 1D10D3 1D10D9
CA14004-214CS 345.65 CAD
CA14004-214 CA16200-110 CA16200-114 CA89128-816 77507-006
CIP 100® Alkaline Process and Research Cleaners
Cleaning Agents Detergents and Surface Cleansers
CIP 100® detergent is a proprietary blend of potassium hydroxide, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in clean-in-place (CIP), clean-out-of-place (COP) and manual applications.
  • Flexibility - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP),
More Product Information