Alcojet® Low-Foaming Powdered Detergents
Supplier: Alconox
1401 1403 1425 1450 1404
21834-045EA 2277.62 CAD
21834-045 21834-055 21834-075 21834-084 21834-051
Alcojet® Low-Foaming Powdered Detergents
Cleaning Agents Detergents and Surface Cleansers
Non-ionic, low foaming powder cleaner for use in labware washers, parts washers, power-spray systems, manual and ultrasonic cleaning. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass. Free rinsing for reliable results and no interfering residues.
  • Concentrated
  • Biodegradable and readily disposable
  • High performance from your cleaning machinery without damaging washer parts
  • Free rinsing to give you reliable results and no interfering residues
  • Dilute 1:200
For removing soil, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, inorganic residues, particulates, solvents, and chemicals.
They can be used to clean healthcare instruments, laboratory ware, sampling apparatus, plate glass, pharmaceutical apparatus, cosmetics manufacturing equipment, restaurant equipment, wine glasses, counters, floors, sidewalks and driveways, metal castings, forgings and stampings, industrial parts, tanks and reactors.
It can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly.
Note: Corrosion testing is advisable.
Cleaning method: Laboratory warewashers, water-sterilizers, washer-sanitizers, cart washers, cage washers, spray washers, electrocleaning, ultrasonic cleaners, flow through clean-in-place. Manual cleaning if proper skin and eye protection is used.
Certifications: Authorized by USDA for use in federally inspected meat and poultry plants. FDA certified.