Citranox® Acid Cleaner and Detergents
Supplier: Alconox
1815 1801 1855
CA21835-353EA 1886.74 CAD
CA21835-353 CA21835-350 CA21835-356
Citranox® Acid Cleaner and Detergents
Cleaning Agents Detergents and Surface Cleansers
CITRANOX is a phosphate free, concentrated cleaner and metal brightener for high performance removal of metal oxides, scale, salts and inorganic residues in manual and ultrasonic cleaning. Corrosion inhibited and free rinsing for reliable results and no interfering residues. Ideal for trace metal analysis.
  • Concentrated to save you money
  • Phosphate free, biodegradable and readily disposable
  • Penetrating wetting power to save you time
  • Free rinsing to give you reliable results and no interfering residues
  • Use to pass your critical cleaning validation tests for lab accreditation and plant inspection approval
  • Dilute 2:100
For removing scale, metal oxides, metal complexes, trace inorganics, milkstone, pasta and starch, soil, grit, buffing compound, grime, grease, fats, oils, particulates, deposits, chemical and solvents.
They can be used to clean airy equipment, laboratory ware, sampling apparatus, lab stills, laser chillers, clean rooms, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, restaurant equipment, rethermolators, steam tables, metal forgings, stampings and castings, industrial parts, pipes, tanks, boilers and reactors.
Cleaning method: Soak, brush, sponge, cloth, ultrasonic cleaner, flow through clean-inplace.
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, bronze, aluminum, and zinc if rinsed promptly.
Note: Corrosion testing is advisable.
Certifications: Authorized by USDA for use in federally inspected meat and poultry plants. FDA certified.
Caution: Will foam—not for spray or machine use.