CIP 200® Acid Cleaner, STERIS®
Supplier: Steris
1D2002 1D2001 1D2008
CA14004-132EA 2310.11 CAD
CA14004-132 CA14004-134 CA16200-116 CA95000-008
CIP 200® Acid Cleaner, STERIS®
Cleaning Agents Detergents and Surface Cleansers
CIP 200® is a high performance, phosphoric acid cleaner for use in manual, immersion, and recirculating spray equipment including clean-in-place (CIP) systems.
  • Flexibility - Effective cleaning against a wide variety of pharmaceutical process residues CIP, clean-out-of-place (COP), and manual applications
  • Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Cost Savings - Reduces water consumption and cleaning time due to
More Product Information