Corning® Coliform Water Test Sample Container, Corning
Supplier: Corning
1705-100 1700-100
27614-453CS 155.72 CAD
27614-453 66175-105
Corning® Coliform Water Test Sample Container, Corning
Vials
These graduated, sterile, cylindrical vials made from PP are used in testing for the presence of coliform bacteria, microbiological contaminants in drinking water.
  • One-piece container with attached lid to reduce chance of contamination
  • Locking arrow ensures sterility
  • Leak tight when sealed properly
  • Tie-down to protect from accidental opening
  • With or without sodium thiosulfate tablet
Meets EPA requirements
Ordering information: Includes 100 containers per case. Comes with sample labels and instructions.