MICRO-90® Concentrated Cleaning Solution, International Products
Supplier: International Products
M-9033 M-9034-055 M-9050-12 M-9051-04
21830-420EA 1327.91 CAD
21830-420 21830-422 89210-138 89210-140
MICRO-90® Concentrated Cleaning Solution, International Products
Cleaning Agents Detergents and Surface Cleansers
A powerful, multi-purpose liquid for precision cleaning of glassware, filter membranes, laboratory equipment, electronics, metal parts, plastics, and other surfaces
  • Mildly alkaline, aqueous cleaner lifts, disperses, emulsifies, sequesters, dissolves, suspends, and decomposes soils
  • Concentrate mixes instantly with water
  • Safe to use
  • Cleaner is environmentally friendly, with no CFCs, ODCs, phosphates, or phenols
Highly-concentrated — a one liter container of cleaner makes over 12 gallons at 2% solution.
Best used in manual, immersion, and low-pressure or low-agitation systems. Ideal for use in ultrasonic cleaners.
Certifications: Cleaner is NSF-registered as an A1, general cleaner in and around food-processing areas and equipment.
Ordering information: Validation methods are available upon request.