VWR® TraceClean® 60 ml Vials
Supplier: VWR International
89093-858 89093-864 89093-872 89093-876 89093-868
89093-858CS 614.26 CAD
89093-858 89093-864 89093-872 89093-876 89093-868
VWR® TraceClean® 60 ml Vials
Vials
Type I clear borosilicate glass vials are designed for use in various analyses and in specimen, extract, and standards storage.
  • Borosilicate glass fabrication
  • Supplied with pen-top caps
  • Prepared to avoid volatile contamination
Vials are prepared in a solvent-free facility to prevent possible volatile contamination
Processed (PC) and Quality-Assured® (QA) vials follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA vials include a Certificate of Analysis inMore Product Information