Cage-Klenz® 150 Alkaline Detergents
Supplier: Steris
1K1505 1K1502 1K15D3
CA14004-226EA 452.21 CAD
CA14004-226 CA14004-228 77531-216
Cage-Klenz® 150 Alkaline Detergents
Cleaning Agents Detergents and Surface Cleansers
Cage-Klenz® 150 Detergent is a liquid, phosphate-free chlorinated alkaline detergent for use in high pressure spray animal cage washing equipment. This highly alkaline, oxidative chemistry is very effective at removing stubborn organic residues.
  • Efficacy - Chlorinated alkaline formula effectively cleans animal fats, oils and other organic cage residues in both soft and hard water
  • Flexibility - Low foaming for use in automated spray washing applications
  • Cost Savings - Reduces utility consumption and cleaning time with free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Cage-Klenz® 150 Detergent is specially formulated to meet the most challenging cleaning and materials compatibility requirements found in research animal caging.