Precleaned VOA Vials, Environmental Express®
Supplier: Environmental Express
APC1590P APC1675P APC3675P APC3590P APC3650 APC1601 APC1560 APC1618 APC1628 APC1624 APC3550P APC3660P APC1556 APC1541 APC1598 APC1596 APC1630 APC1545 APC1559 APC1553P
23050-008CS 632.76 CAD
23050-008 23050-010 23050-054 23050-078 23050-116 23050-298 23050-340 23050-378 23050-384 23050-386 23050-026 23050-030 89202-862 89202-866 89202-870 89202-872 89202-874 89202-876 89202-882 89202-884
Precleaned VOA Vials, Environmental Express®
Vials
These volatile organic analysis (VOA) vials are made of borosilicate glass. Choose from an open-top cap (0.125" thick PTFE/silicone septa) or closed-top style cap in a variarty of capacities and clear or amber glass.
Preserved vials come pre-filled with common preservative reagents for convenience and safety.
Certifications: All containers meet one of two levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level I indicates that each case of containers has been custody-sealed, lot number-labeled for traceability, and a Certificate of Analysis indicating that the containers received full EPA quality assurance treatment.
Accessories information: The accessory Foam Block Rack holds 72×40 ml vials. Ideal for storage and shipping 40 ml VOA vials.