Synthware Distallation Apparatus, Vacuum, Kemtech America
Supplier: Kemtech America
D101410 D101910 D101414
60002-116EA 526.02 CAD
60002-116 60002-118 60002-120
Synthware Distallation Apparatus, Vacuum, Kemtech America
Distillation Components Distillation Sets
Joint on side tube accommodates 25 mm immersion thermometer.
  • Heavy wall Borosilicate construction
  • Ground glass joint
  • Jacketed
  • Designated 10/18 Thermometer joint
10/18 joint on side tube accommodates 25 mm immersion thermometer.
Ordering information: Thermometer is not supplied; contact your Avantor representative for more information.