VWR® Jars, PTFE
Supplier: VWR International
89026-156 89026-158 89026-160 89026-164 89026-166 89026-168 89026-170 89026-172
89026-156EA 1175 CAD
89026-156 89026-158 89026-160 89026-164 89026-166 89026-168 89026-170 89026-172
VWR® Jars, PTFE
Jars General Use Jars
Isostatically molded, pure FDA approved PTFE molded stackable jars have thick wall construction and a smooth internal finish.
  • Chemically resistant and inert
  • Screw-on cap included
Suitable for IPA Method 1311 TCLP.
Caution: Do not use with cap sealed for chemical reactions or digestions. Loosen lid when thermally sterilizing.