VWR® TraceClean® 40 ml Vials
Supplier: VWR International
89093-832 89093-836 89093-842 89093-844 89093-846 89093-850 89093-862 89093-870 89093-874 89093-866
89093-832CS 426.85 CAD
89093-832 89093-836 89093-842 89093-844 89093-846 89093-850 89093-862 89093-870 89093-874 89093-866
VWR® TraceClean® 40 ml Vials
Vials
Type I borosilicate glass vials are for use in volatile organic analyses and are prepared in a solvent-free facility to prevent possible volatile contamination.
  • Uniform 40 ml capacity
  • Clear and amber formats available
  • Custody-sealed packaging to ensure content integrity
Vials are available with polypropylene open-top or solid-top caps.
Processed (PC) and Quality-Assured® (QA) vials follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA vials include aMore Product Information