VWR® TraceClean® 20 ml Vials
Supplier: VWR International
89093-830 89093-838 89093-848 89093-854 89093-860 89096-774
89093-830CS 388.54 CAD
89093-830 89093-838 89093-848 89093-854 89093-860 89096-774
VWR® TraceClean® 20 ml Vials
Vials
Type I clear borosilicate glass vials are for use in various analyses and in specimen, extract, and standards storage.
  • Borosilicate glass fabrication
  • Available with polypropylene solid-top or open-top caps
  • Prepared to avoid volatile contamination
They are prepared in a solvent-free facility to prevent possible volatile contamination
Processed (PC) and Quality-Assured® (QA) vials follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA vials include a CertificateMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable