EPA Vials, Borosilicate Glass, with PTFE/Silicone-Lined Cap, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
60961B-912 60961C-4 60961C-6 60961C-912
66014-295CS 2568.14 CAD
66014-295 66009-977 66014-303 66014-304 66014-298
EPA Vials, Borosilicate Glass, with PTFE/Silicone-Lined Cap, Kimble Chase
Vials
For the collection and storage of water samples.
  • Made from KG-33 clear borosilicate glass
Clear glass vials have 0.005" PTFE-faced 0.120" silicone rubber septa with open-top screw-caps attached.
Comply with guidelines for establishing test procedures for the analysis of pollutants (Ref. EPA Methods 601, 602, 603, and 604).
Ordering information: Vials are packaged after the production process in shrink-wrapped modules to help reduce particulate contamination.