Nalgene® Serum Vials, Crimp Finish, PETG, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
342030-0003 342030-0005 342030-0020
13500-106CS 2609.56 CAD
13500-106 13500-108 13500-112
Nalgene® Serum Vials, Crimp Finish, PETG, Sterile, Thermo Scientific
Vials
Break-resistant vials are ideal for packaging diagnostic serum, biological serum, veterinary pharmaceuticals, and ophthalmic pharmaceuticals.
  • Non-cytotoxic
  • Non-pyrogenic
  • Made of glass-clear PETG
Manufactured and packaged in controlled environment HEPA-filtered work areas. Sterilized to 10⁻⁶ sterility assurance level.
Certifications: Vials meet USP Class VI and USP 661 standards. Suitable for air shipment; quality tested to ensure leakproof performance.
Ordering information: Vials are packaged in trayless shrink-wrap modules. Rubber stoppers and crimp caps not included.