Synthware Conical Reaction Vials, Graduated, with 14/20 Joint, Microscale, Kemtech America
Supplier: Kemtech America
V141003 V141005 V141008
60004-176EA 32 CAD
60004-176 60004-178 60004-180
Synthware Conical Reaction Vials, Graduated, with 14/20 Joint, Microscale, Kemtech America
Vials
Graduated, heavy wall reaction vials have an internal conical bottom and external flat bottom.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Graduated
Suitable for microscale applications.
[ST] Joint No: 14/20.