Synthware Conical Reaction Vials, Heavy Wall
Supplier: Kemtech America
V130501 V130701 V130711 V131003 V131005 V131008
60004-162EA 25.49 CAD
60004-162 60004-164 60004-166 60004-168 60004-170 60004-172
Synthware Conical Reaction Vials, Heavy Wall
Vials
Graduated.[ST] Joint No.: 14/10.
  • Heavy wall Borosilicate construction
  • Threaded Ground glass joint
  • Graduated
  • Cap included
This heavy wall reaction vial is great for microscale reactions.
Ordering information: For replacement caps, Viton® O-rings, or septa, contact your Avantor representative.
Delivery information: Supplied with one compression cap, one Viton® O-ring, and one fluoropolymer resin-faced silicone rubber septum.