E-Z Ex-Traction™ High Recovery Vials, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W224613 W224601 W224603 W224617 W224618 W224619 W224630 W224632 W224634 W224636
80076-564CS 1414.69 CAD
80076-564 89042-296 89042-298 89042-300 89042-302 89042-304 89042-306 89042-308 89042-310 89042-312
E-Z Ex-Traction™ High Recovery Vials, Wheaton, DWK Life Sciences
Vials
Precision-manufactured from low extractable borosilicate glass, these vials are ideal for compound management applications.
A variety of caps are available for these high recovery vials: black polypropylene open-top caps with fluoropolymer resin-faced silicone rubber liner, black phenolic screw caps with fluoropolymer resin-faced (14B) rubber liner, and closed and open-top white polypropylene screw caps with bonded fluoropolymer resin-faced silicone liners.
The conical well design ensures maximum recovery of high value samples. Vials feature a centered conical apex, which is important for automated applications. The large, smooth, and flat bottom provides stability as well as ample area for large bar codes.
Certifications: Vials meet ASTM Type I, Class A and USP Type I standards.