WHEATON® ABC Screw Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W225330-01 W225332-0201 W225332-0203 W225332-0204 W225332-0205 W225332-0207 W225333-03 W225333-0301 W225336-06 W225336-0601 W225338-08 W225338-0801 W225330-0101 W225330-0103 W225330-0104 W225330-0105 W225330-0107 W225332-02 W225338-0804 W225334A-04 W225334A-0401
14213-300CS 638.83 CAD
14213-300 14213-314 14213-316 14213-318 14213-320 14213-322 14213-324 14213-326 14213-332 14213-334 14213-336 14213-338 89232-020 89232-022 89232-024 89232-026 89232-028 89235-506 89235-508 89235-510 89235-512
WHEATON® ABC Screw Caps, DWK Life Sciences
Closures Chromatography Vial Closures
These closures for ABC Vials feature Wheaton Super Septa and low extractable septa.
  • Manufactured from polypropylene
  • 100/pack, 10 packs/case
Available in natural, black, red, blue, yellow or green polypropylene