WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W225151 W225150-02 W225150-0201 W225150-0204 W225150-0205 W225150-0207 W225150-03 W225150-0301 W225150-06 W225150-0601 W225150-08 W225150-0801 W225151-02 W225151-0201 W225151-0204 W225151-0205 W225151-0207 W225151-0301 W225151-06 W225151-0601 W225151-08 W225151-0801 W225152-02 W225152-0204 W225152-0205 W225152-0207 W225152-03 W225152-0301 W225152-04 W225152-06 W225152-0601 W225152-08 W225152-0801 W225153-02 W225153-0201 W225153-0204 W225153-0205 W225153-0207 W225153-03 W225153-0301 W225153-06 W225153-0601 W225153-08 W225153-0801 W225154-04 W225154-0401 W225156-04 W225156-0401 W225157-04 W225157-0401 W225154-01 W225154-0103 W225154-0104 W225154-0105 W225154-0107 W225155-01 W225155-0101 W225155-0104 W225155-0105 W225155-0107 W225156-01 W225156-0101 W225156-0103 W225156-0104 W225156-0107 W225157-01 W225157-0101 W225157-0103 W225157-0104 W225157-0105 W225157-0107 W225150 W225152 W225153 W225150-0804 W225151-0804 W225152-0804 W225153-0804
14213-342CS 739.76 CAD
14213-342 14213-456 14213-458 14213-462 14213-464 14213-466 14213-468 14213-470 14213-476 14213-478 14213-480 14213-482 14213-496 14213-498 14213-502 14213-504 14213-506 14213-510 14213-516 14213-518 14213-520 14213-522 14213-536 14213-538 14213-542 14213-544 14213-546 14213-548 14213-550 14213-552 14213-556 14213-558 14213-560 14213-562 14213-576 14213-578 14213-582 14213-584 14213-586 14213-588 14213-590 14213-596 14213-598 14213-600 14213-602 14230-520 14230-522 14230-528 14230-530 14230-532 14230-534 89232-030 89232-032 89232-034 89232-036 89232-038 89232-040 89232-042 89232-044 89232-046 89232-048 89232-050 89232-052 89232-054 89232-056 89232-058 89232-060 89232-062 89232-064 89232-066 89232-068 89232-070 89232-072 89232-074 89232-076 89235-494 89235-496 89235-498 89495-278 89495-280 89495-282 89495-284
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Vials
ABC Vials™ are constructed of Type 1 borosilicate glass.
  • 40% wider opening than standard screw-top vials
  • 9 mm PP ABC screw-thread caps are available in six liner types
ABC Vials™ feature a superior thread design that provides a more secure seal to the closure. The closures feature Wheaton Super Septa," a low extractable septa that is optimal for GC/MS applications.
Ordering information: Available with or without writing patch.
Delivery information: Convenience packs include 1.8 ml vials packaged in a recyclable PETE plastic clamshell and caps inMore Product Information