WHEATON® LVI™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W225326-02 W225326-0201 W225326-0204 W225326-0205 W225326-0207 W225326-03 W225326-0301 W225326-04 W225326-0401 W225326-06 W225326-0601 W225326-08 W225326-0801 W225328-02 W225328-0201 W225328-0204 W225328-0205 W225328-0207 W225328-03 W225328-0301 W225328-04 W225328-0401 W225328-06 W225328-0601 W225328-08 W225328-0801 225326 225328 W225327-01 W225327-0101 W225327-0103 W225327-0104 W225327-0105 W225327-0107 W225329-01 W225329-0101 W225329-0103 W225329-0104 W225329-0105 W225329-0107
14213-896CS 448.01 CAD
14213-896 14213-898 14213-902 14213-904 14213-906 14213-908 14213-910 14213-912 14213-914 14213-916 14213-918 14213-920 14213-922 14213-936 14213-938 14213-942 14213-944 14213-946 14213-948 14213-950 14213-952 14213-954 14213-956 14213-958 14213-960 14213-962 16150-410 16150-412 89232-078 89232-080 89232-082 89232-084 89232-086 89232-088 89232-090 89232-092 89232-094 89232-096 89232-098 89232-100
WHEATON® LVI™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Vials
These large-opening vials feature a sealed-in, limited-volume insert which allows more consisent sample recovery from vial to vial and reduces the possibility of autosampler needle damage.
Inserts and clear vials are manufactured from ASTM Type I, Class A borosilicate glass. Amber vials are manufactured from ASTM Type I, Class B borosilicate glass. All LVI™ vials include a white ceramic write-on patch for easier sample identification and are compatible with all autosamplers that use 12×32 mm vials.
The insert has a slight flare at the top that provides a larger needle target and the manufacturing process precisely controls the depth and position of the insert relative to the bottom of the vial. LVI™ vials have a 300 µl capacity with a residual volume of only 4 µl, depending on the solvent, needle style, and depth adjustment of the autosampler.
Ordering information: For LVI™ vials with 11 mm crimp-top finish, see 66014-568 series.