WHEATON® E-Z Vial® Large-Opening Crimp-Top Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
225174 W225175-0103 W225175-0104 W225175-0107 W225175-02 W225175-0203 W225175-0204 W225175-0207 W225175-0303 W225175-0304 W225174-0103 W225174-0104 W225174-0107 W225174-02 W225174-0203 W225174-0204 W225174-0207 W225174-0303 W225174-0304 W225172-01 W225172-0103 W225172-0104 W225172-0107 W225172-02 W225172-0203 W225172-0204 W225172-0207 W225172-0303 W225172-0304 W225173-0103 W225173-0104 W225173-0107 W225173-02 W225173-0203 W225173-0204 W225173-0207 W225173-0303 W225173-0304 225175 225172 225173
66014-534CS 711.71 CAD
66014-534 14217-406 14217-408 14217-410 14217-412 14217-414 14217-416 14217-418 14217-422 14217-424 14217-438 14217-440 14217-442 14217-444 14217-446 14217-448 14217-450 14217-454 14217-456 14217-468 14217-470 14217-472 14217-474 14217-476 14217-478 14217-480 14217-482 14217-486 14217-488 14217-502 14217-504 14217-506 14217-508 14217-510 14217-512 14217-514 14217-518 14217-520 89235-514 89235-516 89235-518
WHEATON® E-Z Vial® Large-Opening Crimp-Top Vials, DWK Life Sciences
Vials
Feature a 40% larger neck opening than standard opening vials, which improves sample accessibility while reducing autosampler needle damage.
Vials measure 12×32 mm with a 1.8 ml volume and 11 mm crimp-top. Seals feature 5 ml septa bonded to another material.
Ordering information: Vials are packaged in shrink-wrapped trays of 100. Aluminum seals are packaged in plastic bags. Glass and polypropylene limited volume inserts are also available separately; contact your Avantor sales representative.